Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 4/ANGPTL4, Mouse anti-Human, Clone: Kairos-1, Novus Biologicals™

Mouse Monoclonal Antibody

$261.50 - $345.50

Specifications

Antigen Angiopoietin-like Protein 4/ANGPTL4
Clone Kairos-1
Applications Western Blot, ELISA, Immunohistochemistry
Host Species Mouse
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB013155
View Documents
Novus Biologicals
NBP280039100UG
100 ug
Each for $345.50
Add to cart
 
NB013156
View Documents
Novus Biologicals
NBP28003950UG
50 ug
Each for $261.50
Add to cart
 
Description

Description

Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human Samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications

Specifications

Angiopoietin-like Protein 4/ANGPTL4
Western Blot, ELISA, Immunohistochemistry
Human
Primary
Kairos-1
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit