Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 4/ANGPTL4, Mouse anti-Human, Clone: Kairos4-397G, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP280042100UG

 View more versions of this product

Catalog No. NB013161


Add to cart

Description

Description

Angiopoietin-like Protein 4/ANGPTL4 Monoclonal specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Angiopoietin-like Protein 4/ANGPTL4
Kairos4-397G
Unconjugated
0.2 um-filtered solution in PBS, pH 7.4
angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein
Mouse
IgG1 Kappa
100 ug
Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
Primary
51129
Human
Western Blot, ELISA, Immunohistochemistry
1 mg/mL
Western Blot, ELISA, Immunohistochemistry
Purified
ANGPTL4
Recombinant human coiled-coil domain of Angiopoietin-like Protein 4/ANGPTL4.
Protein G purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Recognizes the coiled-coil domain (CCD) of human Angiopoietin-like Protein 4/ANGPTL4. Detects a band of ∽19kDa by Western blot.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit