Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiopoietin-like Protein 4/ANGPTL4, Mouse anti-Human, Clone: Kairos4-397G, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP28004250UG
Description
Angiopoietin-like Protein 4/ANGPTL4 Monoclonal specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Angiopoietin-like Protein 4/ANGPTL4 | |
Monoclonal | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry | |
angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Recognizes the coiled-coil domain (CCD) of human Angiopoietin-like Protein 4/ANGPTL4. Detects a band of ∽19kDa by Western blot. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
Kairos4-397G | |
Unconjugated | |
0.2 um-filtered solution in PBS, pH 7.4 | |
ANGPTL4 | |
Recombinant human coiled-coil domain of Angiopoietin-like Protein 4/ANGPTL4. | |
50 μg | |
Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators | |
51129 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title