Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 4/ANGPTL4, Rabbit anti-Human,Mouse, Janelia Fluor 549, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP219016JF549

Catalog No. NB004936

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Angiopoietin-like Protein 4/ANGPTL4 Polyclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Angiopoietin-like Protein 4/ANGPTL4
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein
Rabbit
IgG
0.1 mL
Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
Primary
Human, Mouse
Western Blot, Immunocytochemistry, Immunofluorescence
Western Blot, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
ANGPTL4
Synthetic peptide made to an internal sequence of human ANGPTL4 (within residues 150-250) [UniProt Q9BY76].
Immunogen affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit