Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 7/ANGPTL7 Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0093-9H17, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212425AF750

Catalog No. NP21242A70


Add to cart

Description

Description

Angiopoietin-like Protein 7/ANGPTL7 Monoclonal antibody specifically detects Angiopoietin-like Protein 7/ANGPTL7 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Angiopoietin-like Protein 7/ANGPTL7
MM0093-9H17
Western Blot
Conjugated, Purified
ANGPTL7
Human
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
angiopoietin-like 7, Angiopoietin-like factor, Angiopoietin-like protein 7, angiopoietin-related protein 7, AngX, CDT6angiopoietin-like factor (CDT6), Cornea-derived transcript 6 protein, dJ647M16.1, RP4-647M16.2
Mouse
IgG1
0.1 mL
Primary
10218.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit