Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiopoietin-like Protein 7/ANGPTL7 Mouse anti-Human, Clone: MM0093-9H17, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Angiopoietin-like Protein 7/ANGPTL7 |
---|---|
Clone | MM0093-9H17 |
Concentration | LYOPH |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP212425
|
Novus Biologicals
NBP212425 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB019210
|
Novus Biologicals
NBP2124250.025MG |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Angiopoietin-like Protein 7/ANGPTL7 Monoclonal antibody specifically detects Angiopoietin-like Protein 7/ANGPTL7 in Human samples. It is validated for Western Blot.Specifications
Angiopoietin-like Protein 7/ANGPTL7 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
10218 | |
Human | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MM0093-9H17 | |
Western Blot 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
angiopoietin-like 7, Angiopoietin-like factor, Angiopoietin-like protein 7, angiopoietin-related protein 7, AngX, CDT6angiopoietin-like factor (CDT6), Cornea-derived transcript 6 protein, dJ647M16.1, RP4-647M16.2 | |
ANGPTL7 | |
IgG1 | |
Protein G purified |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title