Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiostatin Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0094-6F34, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212042AF350
Description
Angiostatin Monoclonal specifically detects Angiostatin in Human samples. It is validated for Western Blot.Specifications
Angiostatin | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein G purified | |
Biologically Active Proteins | |
5340.0 | |
Store at 4°C in the dark. |
Western Blot | |
MM0094-6F34 | |
Western Blot | |
PLG | |
Human recombinant Angiostatin | |
0.1 mL | |
Primary | |
Human | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title