Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiotensin Converting Enzyme 2 (ACE2) Polyclonal Antibody, QED Bioscience™

Rabbit Polyclonal Antibody
Supplier: Leinco Technologies Inc 19506100UL

Description
Angiotensin Converting Enzyme 2 (ACE2) Polyclonal antibody specifically detects Angiotensin Converting Enzyme 2 (ACE2) in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Angiotensin Converting Enzyme 2 (ACE2) | |
Polyclonal | |
Unconjugated | |
ACE2 | |
2010305L05Rik; ACE 2; Ace2; ACEH; ACE-related carboxypeptidase; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; angiotensin I converting enzyme 2; angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; anigotensin I converting enzyme 2; anigotensin-converting enzyme-related carboxypeptidase; metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2; renal angiotensin-converting enzyme 2; UNQ868/PRO1885 | |
Fusion protein corresponding to internal residues of human ACE2. | |
100 μL | |
Primary | |
Human | |
Antibody |
Immunohistochemistry (Paraffin) | |
0.5-1.0 mg/mL | |
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3 | |
Q9BYF1 | |
Rabbit | |
Protein A | |
RUO | |
59272 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction