Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AngiotensinII/III Mouse anti-Human, Janelia Fluor 646, Clone: Ang II E7 (BGN/KA/4L), Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB10062346JF646

Catalog No. NB062346646


Add to cart

Description

Description

Angiotensin II/III Monoclonal specifically detects Angiotensin II/III in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

AngiotensinII/III
Ang II E7 (BGN/KA/4L)
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
AGT
Peptide sequence corresponding to angiotensin II, conjugated to a proprietary molecular weight carrier. Peptide sequence: DRVYIHPF
Protein A or G purified
Lipid and Metabolism
Primary
183
Human, Mouse, Rat
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
AGT angiotensinogen (serpin peptidase inhibitor, clade A, member 8), angiotensin I, angiotensin II, ANHU, SERPINA8
Mouse
IgG2a
0.1mL
Store at 4°C in the dark.
Monoclonal
This product reacts with Angiotensin II and III. Negligible reactivity was observed with Angiotensin I and Angiotensinogen.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit