Learn More
Alfa Aesar™ Angiotensinogen (1-14), Porcine
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66239LB0
 View more versions of this product
Catalog No. AAJ66239LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1759.04
Porcine
Lyophilized solid
RUO
Angiotensinogen (1-14), porcine
HPLC
5mg
Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser
-30° to -10°C