Learn More
Ohaus™ Angle Rotors
Ohaus™ Angle Rotors for use with Frontier 5000 Series Micro Centrifuge (FC5515)
Manufacturer:  Ohaus™ 30130870
 View more versions of this product
Catalog No. 02-106-1044
Add to cart
Description
Description
  • Angle Rotors are available in a variety of different sizes
  • For use with Frontier 5000 Series Centrifuge
Specifications
Specifications
Angle Rotor
Non-sealable
21,953 x g
0.830 kg
45°
Stainless Steel
24 x 1.5/2.0 mL
15,200 rpm
Frontier 5000 Series Micro Centrifuge