Learn More
ANGPTL1 Mouse anti-Human, Clone: OTI1B2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805608
Catalog No. 50-166-7501
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.
Specifications
Specifications
ANGPTL1
Monoclonal
Unconjugated
ANGPTL1
Lyophilized
ANGPTL1
Human recombit protein fragment corresponding to amino acids 24-491 of human ANGPTL1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI1B2
PBS with 8% trehalose and no preservative; pH 7.3
O95841
ANG3, ANGPT3, ARP1, AngY, UNQ162, dJ595C2.2
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9068