Learn More
Abnova™ ANGPTL5 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00253935T02
Catalog No. 89-933-390
Add to cart
Description
Description
Specifications
Specifications
293T
NM_178127.2
pCMV-ANGPTL5 full-length
100 μL
Human
ANGPTL5 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 44.2kDa
Transient overexpression cell lysate was tested with anti-ANGPTL5 antibody (Mfr. No. H00253935-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot