Learn More
Invitrogen™ ANGPTL5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590304
Description
Immunogen sequence: MMSPSQASLL FLNVCIFICG EAVQGNCVHH STDSSVVNIV EDGSNAKDES KSNDTVCKED CEESCDVKTK ITREEKHFMC RNLQNSIVSY TRSTKKLLRN MMDEQQASLD YLSNQVNELM NRVLLLTTEV FRKQLDPFPH RPVQSHGLDC TDIKDTIGSV TKTPSGLYII HPEGSSYPFE VMCDMDYRGG GRTVIQKRID GIIDFQRLWC DYLDGFGDLL GEFWLGLKKI FYIVNQKNTS FMLYVALESE DDTLAYASYD NFWLEDETRF FKMHLGRYSG NAGDAFRGLK KEDNQNAMPF STSDVDNDGC RPACLVNGQS VKSCSHLHNK TGWWFNECGL ANLNGIHHFS GKLLATGIQW GTWTKNNSPV KIKSVSMKIR RMYNPYFK; Positive Samples: SH-SY5Y.
Predicted to be active in collagen-containing extracellular matrix and extracellular space.
Specifications
ANGPTL5 | |
Polyclonal | |
Unconjugated | |
ANGPTL5 | |
angiopoietin like 5; angiopoietin-like 5; angiopoietin-like protein 5; Angiopoietin-related protein 5; ANGPTL5; UNQ5795/PRO19600 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
253935 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.86 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q86XS5 | |
ANGPTL5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-388 of human ANGPTL5 (NP_8352282). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.