Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANGPTL6 Monoclonal Antibody (766544)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA524370
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
May play a role in the wound healing process. May promote epidermal proliferation, remodeling and regeneration. May promote the chemotactic activity of endothelial cells and induce neovascularization. May counteract high-fat diet-induced obesity and related insulin resistance through increased energy expenditure.
Specifications
ANGPTL6 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8R0Z6 | |
ANGPTL6 | |
E. coli-derived recombinant mouse Angiopoietin-like Protein 6/ANGPTL6 Ala25-Leu457. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Western Blot, Immunocytochemistry | |
766544 | |
Unconjugated | |
ANGPTL6 | |
6330404E11Rik; AGF; angiopoietin like 6; angiopoietin-like 6; angiopoietin-like protein 6; Angiopoietin-related growth factor; angiopoietin-related protein 5; Angiopoietin-related protein 6; Angptl6; ARP3; Arp5; UNQ152/PRO178 | |
Rat | |
Protein A/G | |
RUO | |
70726 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction