Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anhydrous Citric Acid, Powder, USP, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
77-92-9
C6H8O7
192.12
KRKNYBCHXYNGOX-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid
OC(=O)CC(O)(CC(O)=O)C(O)=O
77-92-9
C6H8O7
192.12
KRKNYBCHXYNGOX-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid
OC(=O)CC(O)(CC(O)=O)C(O)=O

CI131, 77-92-9, C6H8O7

Spectrum™ Anhydrous Citric Acid, Powder, USP belongs to a class of drugs known as urinary alkalinizers that are used to treat certain metabolic problems (acidosis) caused by kidney disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 99.5 to 100.5%
Ignition Residue 0.1%
Packaging Poly Pail
Sulfate 0.015%
Quantity 2.5 kg