Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anhydrous Dibasic Calcium Phosphate, Powder, USP, 98-103%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7757-93-9
CaHO4P
136.06
FUFJGUQYACFECW-UHFFFAOYSA-L
calcium hydrogen phosphate
[Ca++].OP([O-])([O-])=O
7757-93-9
CaHO4P
136.06
FUFJGUQYACFECW-UHFFFAOYSA-L
calcium hydrogen phosphate
[Ca++].OP([O-])([O-])=O

C1890, 7757-93-9, CaHPO4

Spectrum™ Anhydrous Dibasic Calcium Phosphate, Powder, USP is used as a dietary supplement and as an antacid. It is listed on the FDA′s Generally Recognized As Safe (GRAS) list of substances. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Arsenic (As) 3 μg/g
Chloride 0.25%
Percent Purity 98 to 103%
Packaging Poly Pail
Quantity 2.5 kg