Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anhydrous Lactose, NF, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
63-42-3
C12H22O11
342.30
GUBGYTABKSRVRQ-UHFFFAOYNA-N
2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
63-42-3
C12H22O11
342.30
GUBGYTABKSRVRQ-UHFFFAOYNA-N
2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

LA103, 63-42-3, C12H22O11

Spectrum™ Anhydrous Lactose, NF is used as a tablet diluent in the pharmaceutical industry. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 0.5%
Ignition Residue 0.1%
Packaging Fiber Drum
Quantity 45 kg
Specific Rotation +54.4° to +55.9°
CAS Max % 100%