Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aniline, ≥99.5% (GC), MilliporeSigma™ Supelco™
$32.50 - $105.00
Specifications
CAS | 62-53-3 |
---|---|
Type | Analytical Standard |
Linear Formula | C6H5 NH2 |
MDL Number | MFCD00007629 |
Formula Weight | 93.13 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-102-0635
|
MilliporeSigma Supelco
517881MLF |
1 mL |
Each for $32.50
|
|
|||||
11-101-9679
|
MilliporeSigma Supelco
517885MLF |
5 mL |
Each for $105.00
|
|
|||||
Description
EnvironmentalSpecifications
62-53-3 | |
C6H5 NH2 | |
93.13 | |
Analytical Standard | |
0.7 mmHg (25°C) | |
1.022 g/mL (at 25°C) | |
184°C | |
Neat |
Analytical Standard | |
MFCD00007629 | |
≥99.5% (GC) | |
1139°F | |
3.22 (185°C, vs. air) | |
-6°C | |
70°C |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
RTECSNumber : BW6650000
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title