Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aniline, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

62-53-3
C6H7N
93.129
PAYRUJLWNCNPSJ-UHFFFAOYSA-N
6115
CHEBI:17296
aniline
C1=CC=C(C=C1)N

High quality chemicals for laboratory and specialized industrial use

Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.02kg
Formula Weight 93.13
Quantity 500mL
Flash Point 70.00°C
Chemical Name or Material Aniline
Grade BAKER ANALYZED™ Reagent