Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aniline (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

This product darkens to a reddish-brown color on storage

Packaging Glass Bottle
Quantity 4L

 View more versions of this product

Catalog No. A740I-4


  Check Availability
Add to cart

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

62-53-3
C6H5NH2
MFCD00007629
Aniline
Certified ACS Grade
C6H7N
93.13
Aminobenzene
≥99.0%

Specifications

Vapor Pressure: 0.5mmHg at 20°C
1.021g/cm³
Pass Test
Liquid
Pass Test
0.01% max.
93.13
1547
3.3g/mol
181°C
8.8
76°C
≥99.0%
62-53-3
Pass Test
0.005% max.
C6H7N
C6H5NH2
Glass Bottle
MFCD00007629
Aminobenzene
Light Yellow
−6.2°C
4L
Aniline
Certified ACS Grade

Safety and Handling

SDS & Safety Documents

DANGER!

Emergency Overview
Combustible liquid. Toxic by inhalation, in contact with skin and if swallowed. Risk of serious damage to eyes. May cause an allergic skin reaction. May cause skin and respiratory tract irritation. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Very toxic to aquatic organisms. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:2
Flammability:2
Instability:0

DOTInformation : DOT Class 6.1, : Poison

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit