Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anilinoacetaldehyde diethyl acetal, 98%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
22758-34-5
C12H19NO2
209.289
DHGUGEBXVGPRRD-UHFFFAOYSA-N
anilinoacetaldehyde diethyl acetal,n-2,2-diethoxyethyl aniline,n-2,2-diethoxyethyl benzenamine,n-phenylaminoacetaldehyde diethyl acetal,acmc-1ch3s,2,2-diethoxyethyl phenylamine,n-2,2-diethoxyethyl aniline #,benzenamine,n-2,2-diethoxyethyl,n-phenyl-2-aminoacetaldehyde diethylacetal
519978
N-(2,2-diethoxyethyl)aniline
CCOC(CNC1=CC=CC=C1)OCC
22758-34-5
C12H19NO2
209.289
DHGUGEBXVGPRRD-UHFFFAOYSA-N
anilinoacetaldehyde diethyl acetal,n-2,2-diethoxyethyl aniline,n-2,2-diethoxyethyl benzenamine,n-phenylaminoacetaldehyde diethyl acetal,acmc-1ch3s,2,2-diethoxyethyl phenylamine,n-2,2-diethoxyethyl aniline #,benzenamine,n-2,2-diethoxyethyl,n-phenyl-2-aminoacetaldehyde diethylacetal
519978
N-(2,2-diethoxyethyl)aniline
CCOC(CNC1=CC=CC=C1)OCC
Quantity 1g
Packaging Glass bottle
Assay Percent Range 98%
Formula Weight 209.29
Chemical Name or Material Anilinoacetaldehyde diethyl acetal