Learn More
Medchemexpress LLC Animal-Free IL-18 M 10ug
SDP
Supplier:  Medchemexpress LLC HYP70642AF10UG
Encompass_Preferred
Recombinant mouse interleukin-18 (IL-18) with a C-terminal His tag, produced without animal-derived components and supplied as a lyophilized powder. Expressed in E. coli, the protein runs at approximately 18 kDa by SDS-PAGE and is provided in a 10 μg quantity. Intended for research applications probing cytokine signaling and immune responses; follow the certificate of analysis and SDS for storage and reconstitution instructions.
  • Recombinant mouse IL-18 with C-terminal His tag.
  • Produced without animal-derived components.
  • Expressed in E. coli and purified for research use.
  • Provided as lyophilized powder for stability.
  • Apparent molecular weight ~18 kDa by SDS-PAGE.
  • Supplied in a 10 μg quantity suitable for assay development.
Catalog No. 50-001-85363