Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotano Animal Handling Elbow Length Catch Glove™

Premium grade suede split leather gloves offer protection against bites, scratches, and lacerations when handling medium-sized primates. Designed for trained professionals working in Lab Animal Research (LAR) facilities.

$2782.91 - $6403.86

Specifications

Color Gray
For Use With (Application) Protection Against Bites, Scratches, Punctures, and Lacerations When Handling Medium-Sized Primates
Glove Cuff Style Gauntlet
Grade Premium
Material Suede Leather
View More Specs

Products 9
Catalog Number Mfr. No. Hand Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Hand Quantity Price Quantity & Availability  
17-116-016
View Documents
Rotano
NBAHOFALH20
Left 20 Gloves/Cs
Case for $2,782.91
Add to cart
 
17-116-013
View Documents
Rotano
NBAHOFALH30
Left 30 Gloves/Cs
Case for $3,889.71
Add to cart
 
17-116-010
View Documents
Rotano
NBAHOFALH50
Left 50 Gloves/Cs
Case for $6,403.86
Add to cart
 
17-116-015
View Documents
Rotano
NBAHOFARH20
Right 20 Gloves/Cs
Case for $2,782.91
Add to cart
 
17-116-012
View Documents
Rotano
NBAHOFARH30
Right 30 Gloves/Cs
Case for $3,889.71
Add to cart
 
17-116-009
View Documents
Rotano
NBAHOFARH50
Right 50 Gloves/Cs
Case for $6,403.86
Add to cart
 
17-116-017
View Documents
Rotano
NBAHOFAPR10
Specific 10 Pair/Cs
Case for $2,782.91
Add to cart
 
17-116-014
View Documents
Rotano
NBAHOFAPR15
Specific 15 Pair/Cs
Case for $3,889.71
Add to cart
 
17-116-011
View Documents
Rotano
NBAHOFAPR25
Specific 25 Pair/Cs
Case for $6,403.86
Add to cart
 
Description

Description

  • Protects against bites, scratches, and lacerations when working with medium-sized primates
  • Premium grade suede leather for dexterity, flexibility, and bite protection
  • Full grain hide on the sleeve provides scratch resistance
  • Fleeced poly cotton liner for added insulation, sweat absorption, and comfort
  • Reinforced double layer on thumb and palm for added protection
  • High strength seam construction for additional strength and durability
  • Elbow length design prevents sleeve roll down
  • Exceeds test method EN388: 2016 + A1:2018 6.5 Puncture Resistance levels
Specifications

Specifications

Gray
Gauntlet
Suede Leather
One Size
Keystone
Protection Against Bites, Scratches, Punctures, and Lacerations When Handling Medium-Sized Primates
Premium
Animal Handling Gloves
NBAH-OFA
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit