Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Animal Skeletons
SCI_ED

Real animal skeletons for a 3-D view.

Supplier:  Ranaco Corporation 160.4200.1

 View more versions of this product

Catalog No. S66922


Add to Cart

Description

Description

Meticulously cleaned, degreased and accurately mounted, these real-bone skeletons provide three-dimensional perspective that books, charts and models cannot. Invaluable aid for teaching dissection in the classroom. Skeletons are painstakingly assembled with pins and wires for permanence.

Grass Frog: Rana forreri, mounted with the legs extended between two clear acrylic plates for easy dorsal and ventral viewing; 6 in. (15.2cm) long (extended)
Bullfrog: Rana catesbeiana, articulated and mounted on a wood base with clear acrylic cover; 10 in. (25.4cm) long, extended
Rat: Rattus norvegicus, fully articulated and mounted on a wood base with a clear acrylic cover; 9 in. (22.9cm) long
Rabbit: Oryctolagus cuniculus, articulated and mounted on a wood base with clear acrylic cover. Excellent for carnivore/herbivore comparisons with the cat; 16 in. (40.6cm) long
Cat: Felix domesticus, articulated and mounted on a wood base with clear acrylic cover; 20 in. (50.8cm) long

Specifications

Specifications

Real Bone Skeleton Rabbit
40.6 cm
Wooden Base
16 in.
Real Bone
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit