Learn More
Best Yet Tag Animal Tags
Small aluminum tabs in set numbered 1 to 100 with 100 washers
Manufacturer:  Best Yet Tag BYT EARTAG
Catalog No. 01-337A
Add to cart
Description
Description
  • Insert tag through hole; place washer in back of hole
  • Bend ends of tag back in rivet fashion
  • Leave space for ear growth
Specifications
Specifications
Animal Tag
Washer
Aluminum
Small