Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anisole, 98%, Spectrum™

Click to view available options
Quantity:
5 g
250 g
1 kg
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
100-66-3
100-66-3
Packaging Drum
Quantity 200 L
CAS Max % 1
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.