Learn More
Cayman Chemical Anisomycin, 22862-76-6, 10 mg
SDP
Supplier:  Cayman Chemical 1130810
Encompass_Preferred
Molecular Formula C14H19NO4, Purity ≥98%, Formula Weight 265.3, Synonyms: 2-[(4-methoxyphenyl)methyl]-3-acetate-(2R,3S,4S)-3,4-pyrrolidinediol, Flagecidin, NSC 76712, Wuningmeisu C; A pyrrolidine antibiotic produced by S. griseolus that inhibits protein and DNA synthesis and activates stress-activated protein kinase, MAPK, and other signal transduction pathways.
Catalog No. NC0472038
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.