Learn More
ANK2 Mouse anti-Human, Mouse, Clone: S105-17, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human ANK2.
Manufacturer:  Abnova Corporation MAB12854
Catalog No. 89-933-567
Add to cart
Description
Description
This gene encodes a member of the ankyrin family of proteins that link the integral membrane proteins to the underlying spectrin-actin cytoskeleton. Ankyrins play key roles in activities such as cell motility, activation, proliferation, contact and the maintenance of specialized membrane domains. Most ankyrins are typically composed of three structural domains: an amino-terminal domain containing multiple ankyrin repeats; a central region with a highly conserved spectrin binding domain; and a carboxy-terminal regulatory domain which is the least conserved and subject to variation. The protein encoded by this gene is required for targeting and stability of Na/Ca exchanger 1 in cardiomyocytes. Mutations in this gene cause long QT syndrome 4. Multiple transcript variants encoding different isoforms have been described. [provided by RefSeq]
Specifications
Specifications
ANK2
S105-17
Mouse monoclonal antibody raised against synthetic peptide of human ANK2.
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
DKFZp686M09125/DKFZp686P0948/FLJ38277/LQT4
Mouse
IgG2a
50 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Murine
Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunocytochemistry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
ANK2
A synthetic peptide corresponding to amino acids 203-496 of human ANK2.
Protein G purification
RUO
Primary
287