Learn More
Abnova™ ANKDD1A 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00348094T01
Catalog No. 89-933-398
Add to cart
Description
Description
Specifications
Specifications
293T
BC101275
pCMV-ANKDD1A full-length
100 μL
Human
ANKDD1A 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 46.6kDa
Transient overexpression cell lysate was tested with anti-ANKDD1A antibody (Mfr. No. H00348094-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot