Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKDD1B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549080

Catalog No. PA549080

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ANKDD1B
Polyclonal
PBS with 0.09% sodium azide
A6NHY2
Ankyrin repeat and death domain-containing protein 1B, ANKDD1B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
728780
Western Blot
Unconjugated
ANKDD1B
Liquid
ANKDD1B
KLH conjugated synthetic peptide between 494-520 amino acids from the C-terminal region of human ANKDD1B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit