Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKHD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109388
Description
Immunogen sequence: VNSSKYPSLL LHSQEEKTST ATSKTQTRLE GEVTPNSLST SYKTVSLPLS SPNIKLNLTS PKRGQKREEG WKEVVRRSKK LSVPASVVSR IMGRGGCNIT AIQDVTGAHI DVDKQKDKNG ERMITIRGGT ESTRYAVQLI NALIQDPAKE LEDLIPKNHI RTPASTKSIH ANFSSGVGTT A.
This gene encodes a protein with multiple ankyrin repeat domains and a single KH-domain. Co-transcription of this gene and the neighboring downstream gene (EIF4EBP3) generates a transcript (MASK-BP3) which encodes a fusion protein comprised of the MASK protein sequence for the majority of the protein and a different C-terminus due to an alternate reading frame for the EIF4EBP3 segments.
Specifications
ANKHD1 | |
Polyclonal | |
Unconjugated | |
ANKHD1 | |
1110004O12Rik; 4933432B13Rik; 9130019P20Rik; A530027J04Rik; A630021B20Rik; AA571404; ANKHD1; ankyrin repeat and KH domain containing 1; ankyrin repeat and KH domain-containing protein 1; HIV-1 Vpr-binding ankyrin repeat protein; hMASK; KIAA1085; MASK; MASK1; mFLJ00246; mKIAA1085; Multiple ankyrin repeats single KH domain; multiple ankyrin repeats, single KH-domain homolog; PP2500; VBARP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100529260, 108857 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
0 | |
ANKHD1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1620-1800 of human ANKHD1 (NP_060217.1). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction