Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKHD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550359
Description
The antibody detects endogenous levels of total ANKHD1 protein.
ANKHD1 Polyclonal antibody specifically detects ANKHD1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
ANKHD1 | |
Polyclonal | |
Unconjugated | |
ANKHD1 | |
1110004O12Rik; 4933432B13Rik; 9130019P20Rik; A530027J04Rik; A630021B20Rik; AA571404; ANKHD1; ankyrin repeat and KH domain containing 1; ankyrin repeat and KH domain-containing protein 1; HIV-1 Vpr-binding ankyrin repeat protein; hMASK; KIAA1085; MASK; MASK1; mFLJ00246; mKIAA1085; Multiple ankyrin repeats single KH domain; multiple ankyrin repeats, single KH-domain homolog; PP2500; VBARP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54882 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8IWZ3 | |
ANKHD1 | |
Fusion protein corresponding to residues near the C terminal of human ankyrin repeat and KH domain containing 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction