Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKHD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116602
Description
Cellular location: Cytoplasm Immunogen sequence: VNSSKYPSLL LHSQEEKTST ATSKTQTRLE GEVTPNSLST SYKTVSLPLS SPNIKLNLTS PKRGQKREEG WKEVVRRSKK LSVPASVVSR IMGRGGCNIT AIQDVTGAHI DVDKQKDKNG ERMITIRGGT ESTRYAVQLI NALIQDPAKE LEDLIPKNHI RTPASTKSIH ANFSSGVGTT A.
This gene encodes a protein with multiple ankyrin repeat domains and a single KH-domain. Co-transcription of this gene and the neighboring downstream gene (EIF4EBP3) generates a transcript (MASK-BP3) which encodes a fusion protein comprised of the MASK protein sequence for the majority of the protein and a different C-terminus due to an alternate reading frame for the EIF4EBP3 segments.
Specifications
ANKHD1 | |
Polyclonal | |
Unconjugated | |
ANKHD1 | |
1110004O12Rik; 4933432B13Rik; 9130019P20Rik; A530027J04Rik; A630021B20Rik; AA571404; ANKHD1; ankyrin repeat and KH domain containing 1; ankyrin repeat and KH domain-containing protein 1; HIV-1 Vpr-binding ankyrin repeat protein; hMASK; KIAA1085; MASK; MASK1; mFLJ00246; mKIAA1085; Multiple ankyrin repeats single KH domain; multiple ankyrin repeats, single KH-domain homolog; PP2500; VBARP | |
Rabbit | |
Affinity chromatography | |
RUO | |
54882 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IWZ3 | |
ANKHD1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1620-1800 of human ANKHD1 (NP_0602171). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction