Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKHD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548746

Catalog No. PA548746

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a protein with multiple ankyrin repeat domains and a single KH-domain. Co-transcription of this gene and the neighboring downstream gene (EIF4EBP3) generates a transcript (MASK-BP3) which encodes a fusion protein comprised of the MASK protein sequence for the majority of the protein and a different C-terminus due to an alternate reading frame for the EIF4EBP3 segments.
Specifications

Specifications

ANKHD1
Polyclonal
PBS with 0.09% sodium azide
Q8IWZ3
Ankyrin repeat and KH domain-containing protein 1, HIV-1 Vpr-binding ankyrin repeat protein, Multiple ankyrin repeats single KH domain, hMASK, ANKHD1, KIAA1085, MASK, VBARP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54882
Western Blot
Unconjugated
ANKHD1
Liquid
ANKHD1
KLH conjugated synthetic peptide between 445-474 amino acids from the N-terminal region of human ANKHD1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit