Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKLE2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$436.00

Specifications

Antigen ANKLE2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP170407
View Documents
Novus Biologicals
NBP170407
100 μL
Each of 1 for $436.00
Add to cart
 
Description

Description

ANKLE2 Polyclonal specifically detects ANKLE2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ANKLE2
Polyclonal
Rabbit
Human
ankyrin repeat and LEM domain containing 2, ankyrin repeat and LEM domain-containing protein 2, FLJ22280, FLJ36132, KIAA0692, LEM domain containing 7, LEMD7
ANKLE2
IgG
Affinity Purified
104 kDa
Western Blot
Unconjugated
RUO
PBS and 2% Sucrose with 0.09% Sodium Azide
23141
Synthetic peptides corresponding to KIAA0692 The peptide sequence was selected from the middle region of KIAA0692. Peptide sequence CYSPSDRQSWPSPAVKGRFKSQLPDLSGPHSYSPGRNSVAGSNPAKPGLG.
Primary
Store at -20C. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit