Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRD13D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179611
Description
ANKRD13D Polyclonal specifically detects ANKRD13D in Mouse samples. It is validated for Western Blot.Specifications
ANKRD13D | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ankyrin repeat domain 13 family, member D, ankyrin repeat domain-containing protein 13D, MGC50828 | |
Rabbit | |
57 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Mouse: 86%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_080996 | |
ANKRD13D | |
Synthetic peptide directed towards the middle region of human Ankrd13dThe immunogen for this antibody is Ankrd13d. Peptide sequence RTEHLSDQDKLRNKGGKTPFQSFLGMAQQHSSHTLAPVQQAASPTNPTAI. | |
Affinity purified | |
RUO | |
338692 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction