Learn More
Abnova™ ANKRD17 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00026057T01
Catalog No. 89-933-430
Add to cart
Description
Description
Specifications
Specifications
293T
BC043394
pCMV-ANKRD17 full-length
100 μL
Human
ANKRD17 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 82.72kDa
Transient overexpression cell lysate was tested with anti-ANKRD17 antibody (Mfr. No. H00026057-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot