Learn More
Abnova™ ANKRD20A1 (-SC) 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00084210T02
Catalog No. 89-933-432
Add to cart
Description
Description
Specifications
Specifications
293T
BC146681
pCMV-ANKRD20A1 (-SC) full-length
100 μL
Human
ANKRD20A1 (-SC) 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 10.23kDa
Transient overexpression cell lysate was tested with anti-ANKRD20A1 (-SC) antibody (Mfr. No. H00084210-B02) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot