Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD20A8P Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549369

Catalog No. PA549369

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ANKRD20A8P
Polyclonal
PBS with 0.09% sodium azide
Q5CZ79
Ankyrin repeat domain-containing protein 20B, Ankyrin repeat domain-containing protein 20A pseudogene, ANKRD20A8P, ANKRD20B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
729171
Western Blot
Unconjugated
ANKRD20A8P
Liquid
ANKRD20A8P
KLH conjugated synthetic peptide between 315-348 amino acids from the Central region of human ANKRD20A8P.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit