Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRD40 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548181
Description
Predicted to react with mouse based on sequence homology.
Specifications
ANKRD40 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q6AI12 | |
ANKRD40 | |
KLH conjugated synthetic peptide between 256-284 amino acids from the C-terminal region of human ANKRD40. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
ANKRD40 | |
Ankyrin repeat domain-containing protein 40, ANKRD40 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91369 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction