Learn More
Abnova™ ANKRD5 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00063926T01
Catalog No. 89-933-456
Add to cart
Description
Description
Specifications
Specifications
293T
BC022878
pCMV-ANKRD5 full-length
100 μL
Human
ANKRD5 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 86.6kDa
Transient overexpression cell lysate was tested with anti-ANKRD5 antibody (Mfr. No. H00063926-B03) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot