Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRD53 Mouse anti-Human, Clone: OTI2E4, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA502595
Description
Required for normal progression through mitosis. Involved in chromosome alignment and cytokinesis via regulation of microtubules polymerization.Specifications
ANKRD53 | |
Monoclonal | |
0.63 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8N9V6 | |
ankyrin repeat domain 53 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
79998 |
Flow Cytometry, Western Blot | |
OTI2E4 | |
Unconjugated | |
ANKRD53 | |
Liquid | |
ANKRD53 | |
Human recombit protein fragment corresponding to amino acids 1-300 of human ANKRD53 produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title