Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD53 Mouse anti-Human, Clone: OTI1E1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525470

Catalog No. PIMA525470


Add to cart

Description

Description

Required for normal progression through mitosis. Involved in chromosome alignment and cytokinesis via regulation of microtubules polymerization.
Specifications

Specifications

ANKRD53
Monoclonal
0.58 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N9V6
ankyrin repeat domain-containing protein 53
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79998
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1E1
Unconjugated
ANKRD53
Liquid
ANKRD53
Human recombit protein fragment corresponding to amino acids 1-300 of ANKRD53 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit