Learn More
ANKRD57 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 240331AP
Catalog No. 50-173-4215
Add to cart
Description
Description
CT1 is a member of the 3-oxoacid CoA-transferase gene family. It is a homodimeric mitochondrial matrix enzyme that plays a central role in extrahepatic ketone body catabolism by catalyzing the reversible transfer of coenzyme A from succinyl-CoA to acetoacetate.This gene encodes a member of the 3-oxoacid CoA-transferase gene family. The encoded protein is a homodimeric mitochondrial matrix enzyme that plays a central role in extrahepatic ketone body catabolism by catalyzing the reversible transfer of coenzyme A from succinyl-CoA to acetoacetate. Mutations in this gene are associated with succinyl CoA:3-oxoacid CoA transferase deficiency.This gene encodes a member of the 3-oxoacid CoA-transferase gene family. The encoded protein is a homodimeric mitochondrial matrix enzyme that plays a central role in extrahepatic ketone body catabolism by catalyzing the reversible transfer of coenzyme A from succinyl-CoA to acetoacetate. Mutations in this gene are associated with succinyl CoA:3-oxoacid CoA transferase deficiency.
Specifications
Specifications
ANKRD57
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q53LP3
ANKRD57, ankyrin repeat domain 57, C2orf26
Rabbit
IgG
150 μL
-20°C
Primary
65124
Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
SOWAHC
Liquid
SOWAHC
ANKRD57 Fusion Protein Ag21238
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human