Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD58 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548293

Catalog No. PA548293

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ANKRD58
Polyclonal
PBS with 0.09% sodium azide
A6NJG2
Ankyrin repeat domain-containing protein SOWAHD, Ankyrin repeat domain-containing protein 58, Protein sosondowah homolog D, SOWAHD, ANKRD58
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
347454
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SOWAHD
Liquid
SOWAHD
KLH conjugated synthetic peptide between 196-224 amino acids from the C-terminal region of human ANKRD58.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit