Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548499

Catalog No. PA548499

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Recruits CKI-epsilon to the beta-catenin degradation complex that consists of AXN1 or AXN2 and GSK3-beta and allows efficient phosphorylation of beta-catenin, thereby inhibiting beta-catenin/Tcf signals.
Specifications

Specifications

ANKRD6
Polyclonal
PBS with 0.09% sodium azide
Q9Y2G4
Ankyrin repeat domain-containing protein 6, Diversin, ANKRD6, KIAA0957
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22881
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ANKRD6
Liquid
ANKRD6
KLH conjugated synthetic peptide between 314-343 amino acids from the Central region of human ANKRD6.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit