Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD60 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548534

Catalog No. PA548534

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ANKRD60
Polyclonal
PBS with 0.09% sodium azide
Q9BZ19
Ankyrin repeat domain-containing protein 60, ANKRD60, C20orf86
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
140731
Western Blot
Unconjugated
ANKRD60
Liquid
ANKRD60
KLH conjugated synthetic peptide between 261-289 amino acids from the C-terminal region of human ANKRD60.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit