Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKS1B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548579

Catalog No. PA548579

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine based on sequence homology.

This gene encodes a multi-domain protein that is predomitly expressed in brain and testis. This protein interacts with amyloid beta protein precursor (AbetaPP) and may have a role in normal brain development, and in the pathogenesis of Alzheimer ft.s disease. Expression of this gene has been shown to be elevated in patients with pre-B cell acute lymphocytic leukemia associated with t(1;19) translocation. Alternatively spliced transcript variants encoding different isoforms (some with different subcellular localization, PMID:15004329) have been described for this gene.
Specifications

Specifications

ANKS1B
Polyclonal
PBS with 0.09% sodium azide
Q7Z6G8
Ankyrin repeat and sterile alpha motif domain-containing protein 1B, Amyloid-beta protein intracellular domain-associated protein 1, AIDA-1, E2A-PBX1-associated protein, EB-1, ANKS1B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56899
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ANKS1B
Liquid
ANKS1B
KLH conjugated synthetic peptide between 570-599 amino acids from the Central region of human ANKS1B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit