Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKS1B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen ANKS1B
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126276
View Documents
Novus Biologicals
NBP31068725UL
25 μg
Each for $106.19
Add to cart
 
NB126275
View Documents
Novus Biologicals
NBP310687100UL
100 μg
Each for $320.71
Add to cart
 
Description

Description

ANKS1B Polyclonal antibody specifically detects ANKS1B in Human samples. It is validated for Western Blot
Specifications

Specifications

ANKS1B
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
56899
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Human
AIDA-1AIDA, amyloid-beta precursor protein intracellular domain associated protein 1, Amyloid-beta protein intracellular domain-associated protein 1, ANKS2, ankyrin repeat and sterile alpha motif domain containing 1B, ankyrin repeat and sterile alpha motif domain-containing protein 1B, cajalin 2, cajalin-2, E2A-PBX1-associated protein, EB1, EB-1MGC26087
The immunogen is a synthetic peptide directed towards the C-terminal region of Human ANKS1B (NP_858056). Peptide sequence PSKPIPKPRVSIRKSVQIDPSEQKTLANLPWIVEPGQEAKRGINTKYETT
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit